Tisk…

7-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 95%

Dodavatel: Apollo Scientific

PC909151-1G PC909151-5G
APOSPC909151-1GEA 6220 CZK
APOSPC909151-1G APOSPC909151-5G
7-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 95%
7-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one
Vzorec: C₁₁H₁₂FNO
MW: 193,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040448
CAS číslo: 676116-80-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR