Tisk…

3-(3-Fluorophenyl)-1,2,4-oxadiazole 96%

Dodavatel: Apollo Scientific

PC909148-10G PC909148-1G PC909148-5G
APOSPC909148-10GEA 14290 CZK
APOSPC909148-10G APOSPC909148-1G APOSPC909148-5G
3-(3-Fluorophenyl)-1,2,4-oxadiazole 96%
3-(3-Fluorophenyl)-1,2,4-oxadiazole
Vzorec: C₈H₅FN₂O
Storage Temperature: Ambient
MDL Number: MFCD16875598
CAS číslo: 1262412-46-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR