Tisk…

6-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 98%

Dodavatel: Apollo Scientific

Synonyms: 6-Fluoro-4,4-dimethyl-3,4-dihydroquinolin-2(1H)-one

PC909146-25G PC909146-5G
APOSPC909146-25GEA 24400 CZK
APOSPC909146-25G APOSPC909146-5G
6-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 98%
6-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one
Vzorec: C₁₁H₁₂FNO
MW: 193,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913844
CAS číslo: 395673-46-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR