Tisk…

6-Fluoro-1-methyl-1,2,3-benzotriazole 96%

Dodavatel: Apollo Scientific

PC909142-1G PC909142-5G
APOSPC909142-1GEA 6220 CZK
APOSPC909142-1G APOSPC909142-5G
6-Fluoro-1-methyl-1,2,3-benzotriazole 96%
6-Fluoro-1-methyl-1,2,3-benzotriazole
Vzorec: C₇H₆FN₃
Storage Temperature: Ambient
MDL Number: MFCD25121771
CAS číslo: 1445322-54-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR