Tisk…

5-Fluoro-2-nitro-N-propylaniline 96%

Dodavatel: Apollo Scientific

PC909133-10G PC909133-1G PC909133-5G
APOSPC909133-10GEA 26650 CZK
APOSPC909133-10G APOSPC909133-1G APOSPC909133-5G
5-Fluoro-2-nitro-N-propylaniline 96%
5-Fluoro-2-nitro-N-propylaniline
Vzorec: C₉H₁₁FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16744200
CAS číslo: 951667-98-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR