Tisk…

3-(2-Fluorophenyl)-1,2,4-oxadiazole 97%

Dodavatel: Apollo Scientific

PC909130-1G PC909130-5G
APOSPC909130-1GEA 3810 CZK
APOSPC909130-1G APOSPC909130-5G
3-(2-Fluorophenyl)-1,2,4-oxadiazole 97%
3-(2-Fluorophenyl)-1,2,4-oxadiazole
Vzorec: C₈H₅FN₂O
Storage Temperature: Ambient
MDL Number: MFCD16875565
CAS číslo: 1262412-83-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR