Tisk…

(3-Fluoro-benzoylamino)-acetic acid 97%

Dodavatel: Apollo Scientific

PC909034-1G PC909034-250MG
APOSPC909034-1GEA 10350 CZK
APOSPC909034-1G APOSPC909034-250MG
(3-Fluoro-benzoylamino)-acetic acid 97%
(3-Fluoro-benzoylamino)-acetic acid
Vzorec: C₉H₈FNO₃
Storage Temperature: Ambient
MDL Number: MFCD00479174
CAS číslo: 366-47-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR