Tisk…

N-Methyl-1-(4-(trifluoromethyl)phenyl)methanamine 97%

Dodavatel: Apollo Scientific

PC908954-25G PC908954-5G
APOSPC908954-25GEA 8570 CZK
APOSPC908954-25G APOSPC908954-5G
N-Methyl-1-(4-(trifluoromethyl)phenyl)methanamine 97%
N-Methyl-1-(4-(trifluoromethyl)phenyl)methanamine
Vzorec: C₉H₁₀F₃N
MW: 189,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115411
CAS číslo: 90390-11-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR