2-Fluoro-4-(methylsulphonyl)toluene 98%
Dodavatel: Apollo Scientific
PC908952-1G
PC908952-5G
APOSPC908952-1GEA
4960
CZK
APOSPC908952-1G
APOSPC908952-5G
2-Fluoro-4-(methylsulphonyl)toluene 98%
2-Fluoro-4-(methylsulphonyl)toluene
Vzorec:
C₈H₉FO₂S MW: 188,219 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14585007 CAS číslo: 828270-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...