Tisk…

2,6-Difluorocinnamic aldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: 2,6-Difluorocinnamaldehyde

PC908912-1G
APOSPC908912-1GEA 7380 CZK
APOSPC908912-1G
2,6-Difluorocinnamic aldehyde 95%
2,6-Difluorocinnamic aldehyde
Vzorec: C₉H₆F₂O
MW: 168,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782064
CAS číslo: 117338-43-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR