8-Fluoro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
Dodavatel: Apollo Scientific
PC908874-1G
PC908874-250MG
APOSPC908874-1GEA
16340
CZK
APOSPC908874-1G
APOSPC908874-250MG
8-Fluoro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
8-Fluoro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine
Vzorec:
C₉H₁₁FN₂ MW: 166,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07369954 CAS číslo: 620948-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...