Tisk…

2-Fluoro-4-methylthio-1-nitrobenzene 95%

Dodavatel: Apollo Scientific

PC908872-1G PC908872-250MG
APOSPC908872-1GEA 16960 CZK
APOSPC908872-1G APOSPC908872-250MG
2-Fluoro-4-methylthio-1-nitrobenzene 95%
2-Fluoro-4-methylthio-1-nitrobenzene
Vzorec: C₇H₆FNO₂S
MW: 187,191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18070982
CAS číslo: 1025509-81-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR