7-Fluoro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
Dodavatel: Apollo Scientific
PC908855-1G
PC908855-250MG
APOSPC908855-1GEA
19440
CZK
APOSPC908855-1G
APOSPC908855-250MG
7-Fluoro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
7-Fluoro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine
Vzorec:
C₉H₁₁FN₂ MW: 166,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07369955 CAS číslo: 886366-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...