6-Bromo-2,3,4-trifluorobenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC908777-1G
PC908777-250MG
PC908777-5G
APOSPC908777-1GEA
4700
CZK
APOSPC908777-1G
APOSPC908777-250MG
APOSPC908777-5G
6-Bromo-2,3,4-trifluorobenzoic acid 98%
6-Bromo-2,3,4-trifluorobenzoic acid
Vzorec:
C₇H₂BrF₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09910412 CAS číslo: 530145-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...