N-Ethyl-2,3-difluorobenzenemethanamine 95%
Dodavatel: Apollo Scientific
PC908769-1G
PC908769-5G
APOSPC908769-1GEA
7670
CZK
APOSPC908769-1G
APOSPC908769-5G
N-Ethyl-2,3-difluorobenzenemethanamine 95%
N-Ethyl-2,3-difluorobenzenemethanamine
Vzorec:
C₉H₁₁F₂N MW: 171,187 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12147622 CAS číslo: 1152832-76-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...