Ethyl 3,3-difluoro-1-methylcyclobutane-1-carboxylate
Dodavatel: Apollo Scientific
PC908371-1G
PC908371-500MG
APOSPC908371-1GEA
36040
CZK
APOSPC908371-1G
APOSPC908371-500MG
Ethyl 3,3-difluoro-1-methylcyclobutane-1-carboxylate
Ethyl 3,3-difluoro-1-methylcyclobutane-1-carboxylate
Vzorec:
C₈H₁₂F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD23105921 CAS číslo: 227607-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...