Tisk…

5-Fluoro-7-methyl-1H-indole 98%

Dodavatel: Apollo Scientific
Varování

PC908205-100MG PC908205-250MG
APOSPC908205-100MGEA 1580 CZK
APOSPC908205-100MG APOSPC908205-250MG
5-Fluoro-7-methyl-1H-indole 98%
5-Fluoro-7-methyl-1H-indole
Vzorec: C₉H₈FN
MW: 149,165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11007869
CAS číslo: 1082041-52-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR