Tisk…

5-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95%

Dodavatel: Apollo Scientific
Varování

PC908171-1G PC908171-250MG
APOSPC908171-1GEA 8340 CZK
APOSPC908171-1G APOSPC908171-250MG
5-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95%
5-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one
Vzorec: C₉H₆BrFO
MW: 229,046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18647721
CAS číslo: 866862-25-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR