9-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC908162-100MG
PC908162-250MG
APOSPC908162-100MGEA
2400
CZK
APOSPC908162-100MG
APOSPC908162-250MG
9-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 97%
9-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one
Vzorec:
C₁₀H₁₀FNO MW: 179,191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14584723 CAS číslo: 1151397-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...