Tisk…

Methyl 3-acetyl-4-fluorobenzoate 98%

Dodavatel: Apollo Scientific

PC908031-1G PC908031-250MG PC908031-5G
APOSPC908031-1GEA 9330 CZK
APOSPC908031-1G APOSPC908031-250MG APOSPC908031-5G
Methyl 3-acetyl-4-fluorobenzoate 98%
Methyl 3-acetyl-4-fluorobenzoate
Vzorec: C₁₀H₉FO₃
MW: 196,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26679549
CAS číslo: 198477-82-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR