2,4-Difluoro-6-hydroxybenzonitrile 95%
Dodavatel: Apollo Scientific
PC908020-1G
PC908020-250MG
APOSPC908020-1GEA
22950
CZK
APOSPC908020-1G
APOSPC908020-250MG
2,4-Difluoro-6-hydroxybenzonitrile 95%
2,4-Difluoro-6-hydroxybenzonitrile
Vzorec:
C₇H₃F₂NO MW: 155.102 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18394850 CAS číslo: 1261793-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...