4-Bromo-2-fluoro-6-methylbenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC908006-1G
PC908006-250MG
PC908006-5G
APOSPC908006-1GEA
4570
CZK
APOSPC908006-1G
APOSPC908006-250MG
APOSPC908006-5G
4-Bromo-2-fluoro-6-methylbenzonitrile
4-Bromo-2-fluoro-6-methylbenzonitrile
Vzorec:
C₈H₅BrFN MW: 214,034 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21337355 CAS číslo: 1427438-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...