Tisk…

Methyl 3-fluorobenzoylacetate 95%

Dodavatel: Apollo Scientific

PC907947-1G PC907947-5G
APOSPC907947-1GEA 5390 CZK
APOSPC907947-1G APOSPC907947-5G
Methyl 3-fluorobenzoylacetate 95%
Methyl 3-fluorobenzoylacetate
Vzorec: C₁₀H₉FO₃
MW: 196,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424771
CAS číslo: 260246-17-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR