Tisk…

Ethyl 2-(3-fluorophenyl)acetate 95%

Dodavatel: Apollo Scientific

PC907941-1G PC907941-5G
APOSPC907941-1GEA 8700 CZK
APOSPC907941-1G APOSPC907941-5G
Ethyl 2-(3-fluorophenyl)acetate 95%
Ethyl 2-(3-fluorophenyl)acetate
Vzorec: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11555043
CAS číslo: 587-47-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR