(1R)-1-(3-Fluorophenyl)ethylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(R)-1-(3-Fluorophenyl)ethanamine, (R)-3-Fluoro-α-methylbenzylamine
PC9079-1G
PC9079-250MG
APOSPC9079-1GEA
3110
CZK
APOSPC9079-1G
APOSPC9079-250MG
(1R)-1-(3-Fluorophenyl)ethylamine 97%
(1R)-1-(3-Fluorophenyl)ethylamine
Vzorec:
C₈H₁₀FN MW: 139,17 g/mol Bod vzplanutí: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD06761859 CAS číslo: 761390-58-3 UN: 2735 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...