3-Fluoro-4-(4-methyl-1H-imidazol-1-yl)benzaldehyde 95%
Dodavatel: Apollo Scientific
PC907868-100MG
PC907868-250MG
APOSPC907868-100MGEA
3270
CZK
APOSPC907868-100MG
APOSPC907868-250MG
3-Fluoro-4-(4-methyl-1H-imidazol-1-yl)benzaldehyde 95%
3-Fluoro-4-(4-methyl-1H-imidazol-1-yl)benzaldehyde
Vzorec:
C₁₁H₉FN₂O Storage Temperature: Ambient |
MDL Number:
MFCD13180648 CAS číslo: 937400-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...