5,6-Difluoroquinazolin-4(3H)-one 95%
Dodavatel: Apollo Scientific
PC907768-1G
PC907768-250MG
APOSPC907768-1GEA
8900
CZK
APOSPC907768-1G
APOSPC907768-250MG
5,6-Difluoroquinazolin-4(3H)-one 95%
5,6-Difluoroquinazolin-4(3H)-one
Vzorec:
C₈H₄F₂N₂O MW: 182.127 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24564974 CAS číslo: 933823-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...