Tisk…

Methyl 3-amino-2,5-difluorobenzoate 95%

Dodavatel: Apollo Scientific

PC907708-1G PC907708-250MG
APOSPC907708-1GEA 5460 CZK
APOSPC907708-1G APOSPC907708-250MG
Methyl 3-amino-2,5-difluorobenzoate 95%
Methyl 3-amino-2,5-difluorobenzoate
Vzorec: C₈H₇F₂NO₂
MW: 187,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17276533
CAS číslo: 1186194-15-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR