Tisk…

[(2-Chloro-4-fluorophenyl)methyl](methyl)amine 95%

Dodavatel: Apollo Scientific

PC907591-1G PC907591-250MG
APOSPC907591-1GEA 22520 CZK
APOSPC907591-1G APOSPC907591-250MG
[(2-Chloro-4-fluorophenyl)methyl](methyl)amine 95%
[(2-Chloro-4-fluorophenyl)methyl](methyl)amine
Vzorec: C₈H₉ClFN
Storage Temperature: Ambient
MDL Number: MFCD09814029
CAS číslo: 823188-81-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR