3-Fluoro-5-methoxyisonicotinaldehyde 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC907544-1G
PC907544-250MG
APOSPC907544-1GEA
11810
CZK
APOSPC907544-1G
APOSPC907544-250MG
3-Fluoro-5-methoxyisonicotinaldehyde 95+%
3-Fluoro-5-methoxyisonicotinaldehyde
Vzorec:
C₇H₆FNO₂ MW: 155,126 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18258153 CAS číslo: 1256792-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...