1-Chloro-3-fluoro-2-isocyanatobenzene 95%
Dodavatel: Apollo Scientific
Synonyms:
1-CHloro-3-fluoro-2-isocyanatobenzene
PC907532-1G
PC907532-5G
APOSPC907532-1GEA
4240
CZK
APOSPC907532-1G
APOSPC907532-5G
1-Chloro-3-fluoro-2-isocyanatobenzene 95%
1-Chloro-3-fluoro-2-isocyanatobenzene
May form small amount of sediment
Vzorec:
C₇H₃ClFNO MW: 171.556 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08444547 CAS číslo: 720678-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...