5-Bromo-2,4-difluoropyrimidine 95%
Dodavatel: Apollo Scientific
PC907459-1G
PC907459-250MG
APOSPC907459-1GEA
7740
CZK
APOSPC907459-1G
APOSPC907459-250MG
5-Bromo-2,4-difluoropyrimidine 95%
5-Bromo-2,4-difluoropyrimidine
Vzorec:
C₄HBrF₂N₂ MW: 194,965 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07787386 CAS číslo: 903131-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...