Tisk…

(2-Fluoro-4-nitrophenyl)methanamine 95%

Dodavatel: Apollo Scientific

PC907324-1G PC907324-250MG
APOSPC907324-1GEA 7410 CZK
APOSPC907324-1G APOSPC907324-250MG
(2-Fluoro-4-nitrophenyl)methanamine 95%
(2-Fluoro-4-nitrophenyl)methanamine
Vzorec: C₇H₇FN₂O₂
MW: 170,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194369
CAS číslo: 937783-91-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR