Tisk…

4-Ethyl-3-fluorophenylboronic acid 95%

Dodavatel: Apollo Scientific

PC907267-1G PC907267-250MG
APOSPC907267-1GEA 16140 CZK
APOSPC907267-1G APOSPC907267-250MG
4-Ethyl-3-fluorophenylboronic acid 95%
4-Ethyl-3-fluorophenylboronic acid
Vzorec: C₈H₁₀BFO₂
Storage Temperature: Ambient
MDL Number: MFCD22491298
CAS číslo: 1374302-04-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR