Tisk…

2,5-Difluoro-4-(trifluoromethyl)benzoic acid 99%

Dodavatel: Apollo Scientific

PC907130-250MG PC907130-1G PC907130-5G
APOSPC907130-250MGEA 4770 CZK
APOSPC907130-250MG APOSPC907130-1G APOSPC907130-5G
2,5-Difluoro-4-(trifluoromethyl)benzoic acid 99%
2,5-Difluoro-4-(trifluoromethyl)benzoic acid
Vzorec: C₈H₃F₅O₂
MW: 226,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631618
CAS číslo: 261945-05-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR