Tisk…

3-Fluoro-5-iodopyridin-2-amine 95%

Dodavatel: Apollo Scientific
Varování

PC907017-10G PC907017-1G PC907017-5G PC907017-100G PC907017-25G
APOSPC907017-10GEA 13430 CZK
APOSPC907017-10G APOSPC907017-1G APOSPC907017-5G APOSPC907017-100G APOSPC907017-25G
3-Fluoro-5-iodopyridin-2-amine 95%
3-Fluoro-5-iodopyridin-2-amine
Vzorec: C₅H₄FIN₂
MW: 238,002 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22628097
CAS číslo: 1321612-85-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR