Tisk…

5-Chloro-2-fluoro-4-methylaniline 95%

Dodavatel: Apollo Scientific

PC906963-1G PC906963-250MG
APOSPC906963-1GEA 19110 CZK
APOSPC906963-1G APOSPC906963-250MG
5-Chloro-2-fluoro-4-methylaniline 95%
5-Chloro-2-fluoro-4-methylaniline
Vzorec: C₇H₇ClFN
MW: 159,589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18426015
CAS číslo: 1263275-21-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR