Tisk…

4-Fluorobenzo[d]thiazol-2(3H)-one 95%

Dodavatel: Apollo Scientific

PC906960-1G PC906960-250MG
APOSPC906960-1GEA 13860 CZK
APOSPC906960-1G APOSPC906960-250MG
4-Fluorobenzo[d]thiazol-2(3H)-one 95%
4-Fluorobenzo[d]thiazol-2(3H)-one
Vzorec: C₇H₄FNOS
MW: 169,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743881
CAS číslo: 63754-97-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR