Tisk…

7-Trifluoromethyl-imidazo[1,2-a]pyridine 95%

Dodavatel: Apollo Scientific

PC906729-1G PC906729-250MG
APOSPC906729-1GEA 17990 CZK
APOSPC906729-1G APOSPC906729-250MG
7-Trifluoromethyl-imidazo[1,2-a]pyridine 95%
7-Trifluoromethyl-imidazo[1,2-a]pyridine
Vzorec: C₈H₅F₃N₂
MW: 186,134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11035989
CAS číslo: 944580-91-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR