(2-Chloro-3-fluorophenyl)methanamine 95%
Dodavatel: Apollo Scientific
PC906725-10G
PC906725-25G
PC906725-5G
APOSPC906725-10GEA
7670
CZK
APOSPC906725-10G
APOSPC906725-25G
APOSPC906725-5G
(2-Chloro-3-fluorophenyl)methanamine 95%
(2-Chloro-3-fluorophenyl)methanamine
Vzorec:
C₇H₇ClFN MW: 159,589 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10687077 CAS číslo: 72235-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...