Tisk…

3-(Prop-2-en-1-yl)azetidine trifluoroacetic acid

Dodavatel: Apollo Scientific

PC906156-1G PC906156-250MG PC906156-500MG
APOSPC906156-1GEA 19940 CZK
APOSPC906156-1G APOSPC906156-250MG APOSPC906156-500MG
3-(Prop-2-en-1-yl)azetidine trifluoroacetic acid
3-(Prop-2-en-1-yl)azetidine trifluoroacetic acid
Vzorec: C₆H₁₁N·CF₃CO₂H
MW: 211,182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28166304
CAS číslo: 1630906-83-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR