Tisk…

3-Fluoro-3-methylcyclobutan-1-amine hydrochloride

Dodavatel: Apollo Scientific

PC905939-100MG PC905939-250MG
APOSPC905939-100MGEA 16770 CZK
APOSPC905939-100MG APOSPC905939-250MG
3-Fluoro-3-methylcyclobutan-1-amine hydrochloride
3-Fluoro-3-methylcyclobutan-1-amine hydrochloride
Vzorec: C₅H₁₀FN·ClH
MW: 139,599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27987246
CAS číslo: 1638768-85-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR