(1-Amino-3-fluorocyclobutyl)methanol hydrochloride
Dodavatel: Apollo Scientific
PC905675-1G
PC905675-250MG
PC905675-500MG
APOSPC905675-1GEA
44310
CZK
APOSPC905675-1G
APOSPC905675-250MG
APOSPC905675-500MG
(1-Amino-3-fluorocyclobutyl)methanol hydrochloride
(1-Amino-3-fluorocyclobutyl)methanol hydrochloride
Vzorec:
C₅H₁₀FNO·ClH Storage Temperature: Ambient |
MDL Number:
MFCD28502513 CAS číslo: 1630907-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...