Tisk…

Methyl 2-(3-fluoro-4-methylphenyl)acetate 95%

Dodavatel: Apollo Scientific

PC905558-1G PC905558-5G
APOSPC905558-1GEA 2320 CZK
APOSPC905558-1G APOSPC905558-5G
Methyl 2-(3-fluoro-4-methylphenyl)acetate 95%
Methyl 2-(3-fluoro-4-methylphenyl)acetate
Vzorec: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025313
CAS číslo: 787585-29-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR