Tisk…

Potassium trifluoro(oxan-4-ylmethoxymethyl)boranuide 95%

Dodavatel: Apollo Scientific
Varování

PC905466-1G PC905466-250MG PC905466-5G
APOSPC905466-1GEA 9530 CZK
APOSPC905466-1G APOSPC905466-250MG APOSPC905466-5G
Potassium trifluoro(oxan-4-ylmethoxymethyl)boranuide 95%
Potassium trifluoro(oxan-4-ylmethoxymethyl)boranuide
Vzorec: C₇H₁₃BF₃O₂·K
MW: 236,081 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11505934
CAS číslo: 1350320-55-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR