4-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC905423-100MG
PC905423-250MG
APOSPC905423-100MGEA
1950
CZK
APOSPC905423-100MG
APOSPC905423-250MG
4-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 98%
4-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C₈H₄BrF₃N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD18258885 CAS číslo: 1256824-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...