Tisk…

2-(Chloromethyl)-3-fluoropyridine 95%

Dodavatel: Apollo Scientific

PC905367-1G PC905367-5G
APOSPC905367-1GEA 2530 CZK
APOSPC905367-1G APOSPC905367-5G
2-(Chloromethyl)-3-fluoropyridine 95%
2-(Chloromethyl)-3-fluoropyridine
Vzorec: C₆H₅ClFN
MW: 145,562 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520697
CAS číslo: 149489-32-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR