Tisk…

4-Chloro-7-fluoro-2-methylquinazoline 97%

Dodavatel: Apollo Scientific

PC905284-1G PC905284-5G
APOSPC905284-1GEA 1310 CZK
APOSPC905284-1G APOSPC905284-5G
4-Chloro-7-fluoro-2-methylquinazoline 97%
4-Chloro-7-fluoro-2-methylquinazoline
Vzorec: C₉H₆ClFN₂
Storage Temperature: Ambient
MDL Number: MFCD12024899
CAS číslo: 1206694-32-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR