2,4-Difluoro-5-methoxybenzaldehyde 95%
Dodavatel: Apollo Scientific
PC905253-1G
PC905253-250MG
APOSPC905253-1GEA
3710
CZK
APOSPC905253-1G
APOSPC905253-250MG
2,4-Difluoro-5-methoxybenzaldehyde 95%
2,4-Difluoro-5-methoxybenzaldehyde
Vzorec:
C₈H₆F₂O₂ MW: 172,129 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24539280 CAS číslo: 177034-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...