1-(3-Fluorophenyl)-1H-pyrazole-4-carbaldehyde 98%
Dodavatel: Apollo Scientific
PC905242-100G
PC905242-10G
PC905242-25G
APOSPC905242-100GEA
32670
CZK
APOSPC905242-100G
APOSPC905242-10G
APOSPC905242-25G
1-(3-Fluorophenyl)-1H-pyrazole-4-carbaldehyde 98%
1-(3-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
Vzorec:
C₁₀H₇FN₂O Storage Temperature: Ambient |
MDL Number:
MFCD05864517 CAS číslo: 936940-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...